Order Entry
Belgium
ContactUsLinkComponent
Potassium chloride 1.0 M KCl volumetric solution, Fluka™
Potassium chloride 1.0 M KCl volumetric solution, Fluka™
Catalog # 60121-1L
CAS Number:  
Potassium chloride 1.0 M KCl volumetric solution, Fluka™
Catalog # 60121-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :74.55 g/mol
Density:1 g/cm³
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360