Order Entry
Belgium
ContactUsLinkComponent
 
J.T.Baker®, Chicken Native Albumin (from Egg)
  0440-01
 :  Avantor
 :  
undefined
J.T.Baker®, Chicken Native Albumin (from Egg)
  0440-01
 :  Avantor
 :  

 

  • Pack type:
    White poly bottle, blue tamper evident cap
  • Pk:
    500 g
  • CAS No.:
    9006-59-1
  • Protein/peptide type:
    Native
  • Source:
    Egg
  • Species:
    Chicken
  • Seq:
    03
  • Storage conditions:
    Refrigerator
  • Form:
    Powder
  • Protein/peptide name:
    Albumin