Order Entry
Belgium
ContactUsLinkComponent
Trichoderma viride cellulase
Trichoderma viride cellulase
Leverancier:  Serva
Certificaten
Trichoderma viride cellulase
Leverancier:  Serva

Specificaties

  • CAS No.:
    9012-54-8
  • Species:
    Trichoderma viride
  • MDL No.:
    MFCD00081510
  • Enzyme name:
    Cellulase

Specificaties

Frequently Bought Together