Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride 5 M in aqueous solution RNase-free
Sodium chloride 5 M in aqueous solution RNase-free
Catalog # J60434.AK
CAS Number:  
Sodium chloride 5 M in aqueous solution RNase-free
Catalog # J60434.AK
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

CAS Number:7647-14-5
 :
Formula:NaCl
MDL Number:MFCD00003477
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3