Order Entry
Belgium
ContactUsLinkComponent
Zinc sulphate 100 mM in aqueous solution
Irritant
Zinc sulphate 100 mM in aqueous solution
Catalog # J60963.AE
CAS Number:  
Irritant
Zinc sulphate 100 mM in aqueous solution
Catalog # J60963.AE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS07
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302