Order Entry
Belgium
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (KOCO(CHOH)₂COONa·4H₂O):
    99.0 - 102.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    6.0 - 8.5
  • Chloride (Cl):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.002 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Ammonium (NH₄):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.005 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together