Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Toxic
Digitonin
Certificates
Digitonin

2 Options Available Below

Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • Melting Pt:240 °C
  • Storage Temperature:Ambient
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤6 % (1 g, 105°C)
  • Appearance of solution:
    (2.5% in warm EtOH) clear colorless to faint yellow
  • Residue after ignition:
    ≤0.3 %
  • Specific optical rotation:
    -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 312,00 €
    312,00 €
    1 * 500 mg
       312,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 690,00 €
    1 690,00 €
    1 * 5 g
       1 690,00 €