Order Entry
Belgium
ContactUsLinkComponent
D-(+)-Galactosamine hydrochloride, Calbiochem®
D-(+)-Galactosamine hydrochloride, Calbiochem®
Catalog # 34539-1
CAS Number:  
D-(+)-Galactosamine hydrochloride, Calbiochem®
Catalog # 34539-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.64 g/mol
EINECS:217-198-1
CAS Number:1772-03-8
Formula:C₆H₁₄ClNO₅
MDL Number:MFCD00135830