Order Entry
Belgium
ContactUsLinkComponent
Anion exchange HPLC columns, NUCLEOGEL® Anion I
Anion exchange HPLC columns, NUCLEOGEL® Anion I
Catalog # MANA719543
Supplier:  MACHEREY-NAGEL
undefined
Anion exchange HPLC columns, NUCLEOGEL® Anion I
Catalog # MANA719543
Supplier:  MACHEREY-NAGEL

Specifications

  • Column Format:
    Guard column
  • Ø int.:
    4,00 mm
  • Particle Size:
    10 µm
  • Length:
    21 mm
  • Seq:
    02
  • pH range:
    1 - 14
  • Phase:
    NUCLEOGEL® Anion I
  • Pk:
    2

Specifications

About this item

This strongly basic polymer-based anion exchange column is used for the analysis of inorganic anions. Contrary to silica-based phase the column is also suited for fluoride analysis.

  • Based on stable polymer (pH 1 to 14)
  • Eluent in column 4 mmol/L salicylate buffer, pH 7,8

The Valco type guard column cartridge is used for analytical Valco column with the guard column holder C (MANA719538).