ContactUsLinkComponent
Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Catalog # 115-1473
Supplier:  Deb-STOKO
undefined
Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Catalog # 115-1473
Supplier:  Deb-STOKO
Specifications
  • Packed:
    70 wipes/bucket, 4 buckets/case
  • Description:
    Kresto kwik-wipes®
  • L×W:
    260×300 mm
  • Pk:
    4
Specifications
About this item
These polypropylene wipes are moistened with a cleansing solution that is ideal for fast cleansing and pre-cleansing after contact with heavy-duty contamination, such as waste mineral and lubricating oils, graphite, metallic dust and carbon black.
  • Ideal for workplaces where there is no water available for cleaning hands
  • Slightly roughened surface provides consistent and reliable cleansing power
  • Non soapy wipes help to stop skin swelling
  • Slightly acidic pH helps to maintain the natural acidic protective mantle
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904