Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chains, Maximum Security

Chains, Maximum Security

Supplier:   Brady
Chains, Maximum Security
Supplier:  Brady
Certificates
Chains, Maximum Security
Chains, Maximum Security

2 Options Available Below

Product Details & Documents

These chains are far more secure than steel cables. They are supplied with a fabric sleeve which will protect paintwork and chrome from damage.

Material: Hardened special chain steel

Diameter: 8 or 10 mm

Length: 110 cm

Product Family Options

Product Information

  • Type
    Colour
    Availability
    Price
  • Ø 8 mm
    Black
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 171,00 €
    171,00 €
    1 * 2 ST
       171,00 €

  • Ø 10 mm
    Black
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 261,00 €
    261,00 €
    1 * 2 ST
       261,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.