Boxer shorts, Match
Supplier: FRISTADS KANSAS
About this item
Boxer shorts made of 95% cotton and 5% Lycra.
- Elasticated waist
Weight: 170 g/m²
6 Options Available Below
Product Details & Documents
Boxer shorts made of 95% cotton and 5% Lycra.
- Elasticated waist
Weight: 170 g/m²
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 113-6703
- 113-6702
- 113-6701
- 113-6704
- 113-6699
- 113-6700
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- Fabric:
- Cotton/Lycra
- Cotton/Lycra
- Cotton/Lycra
- Cotton/Lycra
- Cotton/Lycra
- Cotton/Lycra
- Weight:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-6703Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-6702Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-6701Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-6704Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-6699Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-6700Garment applicationGeneral PurposeGenderFor menFabricCotton/LycraWeight170 g/m²
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



