Solid phase extraction cartridges, HyperSep™ SAX
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Strong anion exchange sorbent for extraction of weak acids.
- Extracts negatively charged compounds from both aqueous and non-aqueous solutions
- Ideally suited for the extraction of weak acids, e.g. carboxylic acids
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by selection of buffer
Frequently Bought Together