Solid phase extraction cartridges, HyperSep™ SCX
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
A strong cation exchange sorbent for extraction of charged basic compounds.
- Benzene ring provides potential for nonpolar interactions
- Low pKa value sorbent
Frequently Bought Together