ContactUsLinkComponent
Membrane filter dispenser, Sentino®
Membrane filter dispenser, Sentino®
Catalog # 516-0665
undefined
Membrane filter dispenser, Sentino®
Catalog # 516-0665
Specifications
  • Description:
    Membrane filter dispenser, Sentino®, EU-/UK-plug
  • Pk:
    1
Specifications
About this item
Filter dispensers are ideal for high throughput microbiology testing laboratories looking for more ergonomic, faster ways to handle individual membranes. The Sentino® filter dispenser is designed to aseptically dispense one disc of membrane at the touch of a button.
  • Easy to load without jamming. Guide plates assist loading and keep refill web in alignment during use
  • Soft touch keypad for aseptically dispensing membrane discs
  • Designed to hold 200 membrane discs
  • Compact, portable design with miniaturised refill pack
  • Use plugged into main power source or charge battery for use anywhere. Lights indicate when battery charge is running low
  • Easy to clean
The Sentino® filter dispenser can be used for: Drinking and wastewater analysis, environmental and recreational water monitoring, personal care product manufacturing, beverage monitoring, pharmaceutical and industrial water system monitoring, pharmaceutical raw materials, intermediates and final product.
Delivery: Supplied with power transformer, EU-plug, UK-plug and user guide CD.
Dispenser pack refills
Product Information