Robotic OMIX™ tips for Tomtec Quadra
Supplier: VARIAN
Certificates
About this item
These Tomtec compatible tips contain a slice of monolithic SPE material.This allows for vacuum-free processing and walk-away automation. With hands-free SPE the process becomes much more streamlined and reproducible.
- Fast, uniform flow maximises productivity and reproducibility
- Small monolithic tip delivers low elution volumes, increasing sensitivity and reducing solvent usage
- Vacuum-free processing improves reproducibility and shortens processing times
Frequently Bought Together