Volumetric flasks, class A, white markings, USP batch certificate, BLAUBRAND®
Supplier: Brand
Certificates
About this item
Borosilicate glass 3.3, amber, with amber glass stopper or PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast white enamel
- DE-M marking
- With USP batch certificate, on request also available with USP individual certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042.
Specifications
- Catalog No:
- 612-5423
- 612-4830
- 612-4833
- 612-5421
- 612-5420
- 612-4829
- 612-5424
- 612-5422
- 612-4831
- 612-5419
- 612-4832
- 612-4824
- 612-5428
- 612-5425
- 612-5427
- 612-5426
- 612-4826
- 612-4825
- 612-4828
- 612-4827
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Specifications
Related Information
Certificates
Search by Lot NumberBrand BLAUBRAND® Volumetric Instruments USP Brand Volumetric Measurement in the Laboratory
Product Literature
Technical Note BRAND Volumetric Flasks
Application Notes
Frequently Bought Together