Orders LinkContactUsLinkComponent
Drum funnels
Drum funnels
Supplier:   HIRSCHMANN
Drum funnels
Supplier:  HIRSCHMANN
Certificates
Drum funnels
3 Options Available Below
Product Details & Documents
These large transparent funnels, made of PP, are suitable for filling large amounts of liquid.
  • Fast discharge by steep 60° angle
  • Convenient handle for hanging
  • Good chemical and temperature resistance
Product Family Options
Product Information
  • Funnel O-Ø
    Stem length
    Length
    Availability
    Price
  • 250 mm
    30 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 105,00 €
    105,00 €
    1 * 6 ST
       105,00 €
  • 350 mm
    35 mm
    440 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,30 €
    36,30 €
    1 * 1 ST
       36,30 €
  • 200 mm
    22 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 63,00 €
    63,00 €
    1 * 6 ST
       63,00 €