Measuring beakers with handle
Catalog # HIRS7600380
Supplier: HIRSCHMANN
Supplier #: 7600380
Log in for availability
About this item
PP, highly translucent.
- Good chemical resistance
- Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
- Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products acc. to EC Directive No. 10/2011.
Frequently Bought Together