Order Entry
Belgium
ContactUsLinkComponent
Ethanol ≥70% (v/v), VOLUSOL®
Ethanol ≥70% (v/v), VOLUSOL®
Catalog # VWRS5966.9010
Supplier:  VWR Collection
CAS Number:  
Ethanol ≥70% (v/v), VOLUSOL®
Catalog # VWRS5966.9010
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :46.07 g/mol
ADR:3,III
 :Filtered through a 0.2 µm filter.
EINECS:200-578-6
UN:1170
 :GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568