Orders LinkContactUsLinkComponent
Capillary columns, CP-SimDist
Capillary columns, CP-SimDist
Supplier:   VARIAN
Capillary columns, CP-SimDist
Supplier:  VARIAN
Certificates
Capillary columns, CP-SimDist
4 Options Available Below
Product Details & Documents
CP-SimDist Fused Silica columns are guaranteed for simulated distillation up to C100.
  • Conforms to ASTM Method D 2887 to provide full compliancy
  • High temperature stationary phase for extended column lifetime
  • Low bleed makes quantitation easier
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    CP-SimDist
    0,17 µm
    0,53 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 657,00 €
    657,00 €
    1 * 1 ST
       657,00 €
  • Capillary
    CP-SimDist
    0,17 µm
    0,53 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 585,00 €
    585,00 €
    1 * 1 ST
       585,00 €
  • Capillary
    CP-SimDist
    0,10 µm
    0,32 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 784,00 €
    784,00 €
    1 * 1 ST
       784,00 €
  • Capillary
    CP-SimDist
    0,10 µm
    0,53 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 130,00 €
    1 130,00 €
    1 * 1 ST
       1 130,00 €