SPE columns, CHROMABOND® PCA
CHROMABOND® PCA is a propylcarboxylic acid modified silica cation exchanger for SPE.
- Base material: Silica
- Weakly acidic cation exchanger (WCX)
Recommended for strong cations like cationic peptides (viscotoxines).
- Catalog No:
- MANA730483
- MANA730482
- MANA730484
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
Frequently Bought Together