Multi-channel pipettes, electronic, variable volume, Acura® electro 956
Supplier: Socorex Isba
Certificates
About this item
These pipettes include a microprocessor-controlled motor, an intuitive software and a rechargeable battery pack. Eight models with 8 and 12 channels cover the volume range 0,5 to 350 µl.
- Lightweight and perfect hand fitting
- Sequential tip ejection
- Shaft height adjustment system called Justip™ (4 mm span) - controlled by efficient click-stops - allows a wide selection of tips to tightly fit the nozzle
- Bowed shape ejector head permits effortless, sequential tip ejection
- 360° rotation of multichannel volumetric modules allows selection of the best working position
- Pipetting cycle counter: simple double click gives access to the number of pipetting cycles performed since last zeroing
Delivery: Pipettes (alone) without plug are supplied with battery pack, QC certificate, operating instructions and samples of Qualitips®.
Pipettes with plug (inital packages) are supplied with individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, Qualitips pipette tip samples.
Pipettes with plug (inital packages) are supplied with individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, Qualitips pipette tip samples.
Caution: The pipette alone is supplied with battery pack but without charger, which is required for operation. It is therefore recommended to purchase an inital package at first. Additional pipettes without charger can be purchased afterwards and charged on the existing stand.
Volumetric modules
Product Information
Frequently Bought Together