Orders LinkContactUsLinkComponent
Capillary columns, PLOT
Capillary columns, PLOT
Supplier:   VARIAN
Capillary columns, PLOT
Supplier:  VARIAN
Certificates
Capillary columns, PLOT
Capillary columns, PLOT
10 Options Available Below
Product Details & Documents
Aluminum oxide PLOT columns offer high selectivity for separating ppm levels of C1 to C5 hydrocarbons in a main stream of C1 to C5 hydrocarbons.
  • High analytical capacity improves efficiency
  • No need for sub-ambient cooling simplifies operation
  • Choice of two polarities for a broad range of applications
These columns analyze more compounds in a single run than packed columns, while still delivering higher resolution and faster analysis times. When compared to liquid stationary phases, the CP-Al₂O₃ PLOT column offers increased selectivity and allows all C1 to C5 hydrocarbon isomers to be separated.
Aluminium oxide PLOT columns are deactivated using very small salt crystals, providing a reproducible and stable deactivation up to 200 °C. Depending on the type of deactivation salt, the CP-Al₂O₃ PLOT column will show a particular selectivity. The KCl salt results in a relatively apolar Al₂O₃ surface, while Na₂SO₄ deactivation provides a polar surface. Unsaturated compounds such as ethylene and acetylene (ethyne) are retained longer.
Product Family Options
CP-Aluminium oxide/Sodium sulfate UltiMetal
Product Information
  • Column Format
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • VARICP6968
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    10,00 µm
    0,53 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 900,00 €
    1 900,00 €
    1 * 1 ST
       1 900,00 €
CP-Aluminium oxide/Potassium chloride
Product Information
  • Column Format
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • VARICP7576
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    4,00 µm
    0,25 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 825,00 €
    825,00 €
    1 * 1 ST
       825,00 €
  • VARICP7577
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    4,00 µm
    0,25 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 420,00 €
    1 420,00 €
    1 * 1 ST
       1 420,00 €
  • VARICP7511
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    5,00 µm
    0,32 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €
  • VARICP7515I5
    Temperature limits -100 to 200/200 °C, 5 in cage
    Temperature limits -100 to 200/200 °C, 5 in cage
    Capillary
    5,00 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 720,00 €
    1 720,00 €
    1 * 1 ST
       1 720,00 €
  • VARICP7515
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    5,00 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 540,00 €
    1 540,00 €
    1 * 1 ST
       1 540,00 €
  • VARICP7519
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    5,00 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 887,00 €
    887,00 €
    1 * 1 ST
       887,00 €
  • VARICP7517
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    10,00 µm
    0,53 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 825,00 €
    825,00 €
    1 * 1 ST
       825,00 €
  • VARICP7518
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    10,00 µm
    0,53 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 580,00 €
    1 580,00 €
    1 * 1 ST
       1 580,00 €
CP-Aluminium oxide/Potassium chloride UltiMetal
Product Information
  • Column Format
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • VARICP6918
    Temperature limits -100 to 200/200 °C, 7 in cage
    Temperature limits -100 to 200/200 °C, 7 in cage
    Capillary
    10,00 µm
    0,53 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 900,00 €
    1 900,00 €
    1 * 1 ST
       1 900,00 €