Gel permeation chromatography columns, MesoPore™
Supplier: VARIAN
Certificates
About this item
MesoPore columns have been specifically designed to give optimum results in the analysis of prepolymers, i.e. polymeric materials with a large oligomeric component.
- Full solvent compatibility with no detrimental effect on efficiency
- Low particle size extracts maximum information from the analyte
- No MWD dislocations so the distribution is a faithful representation of the sample
By combining a 3 µm particle size with high pore volume, MesoPore columns give the highest resolution separations for the analysis of low molecular weight polymers, such as prepolymers, resins, polyols and siloxanes.
Frequently Bought Together