Orders LinkContactUsLinkComponent
Gel permeation chromatography columns, PolyPore™
Gel permeation chromatography columns, PolyPore™
Supplier:   VARIAN
Gel permeation chromatography columns, PolyPore™
Supplier:  VARIAN
Certificates
Gel permeation chromatography columns, PolyPore™
3 Options Available Below
Product Details & Documents
PolyPore columns results unrivalled resolution for the analysis of polymers with broad molecular weight distributions.
  • Routine polymer analysis with very high resolution
  • Wide operating range simplifies column choice
  • Low particle size extracts maximum information from the analyte
Product Family Options
Product Information
  • Column Format
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    5 µm
    7,50 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 350,00 €
    2 350,00 €
    1 * 1 ST
       2 350,00 €
  • Guard column
    5 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
  • Guard column
    5 µm
    7,50 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €