Order Entry
Belgium
ContactUsLinkComponent
Standards for photometric test Kits, NANOCONTROL
Standards for photometric test Kits, NANOCONTROL
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Standards for photometric test Kits, NANOCONTROL
Supplier:  MACHEREY-NAGEL

Specifications

  • Hexavalent chromium:
    No
  • Shipping conditions:
    194 g, 120×101×38 mm
  • Shelf life:
    6 months from date of manufacture, 6 weeks after opening
  • Storage temperature:
    2…8 °C (36…46 °F)
  • Description:
    2 standard solutions, 1 spiking solution, 1 evaluation sheet, 1 instruction leaflet

Specifications

Frequently Bought Together