Order Entry
Belgium
ContactUsLinkComponent
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Health
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAH0772-25G
Supplier:  TCI
CAS Number:  
undefined
Health
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAH0772-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Absorbance:
    max. 0.15 (1mol/L、H2O、260nm)
  • Solubility in Water:
    almost transparency
  • Water:
    max. 7.0 %

Specification Test Results

About this item

 :
 :268.3305 g/mol
EINECS:269-990-1
 :GHS08
Storage Temperature:Ambient
CAS Number:68399-78-0
Formula:C₉H₂₀N₂O₅S
MDL Number:MFCD00038351