Silicon standard solution, 10,000 mg/L Si in water standard for ICP
: CPACHEM
Frequently Bought Together
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
VWR® Economy Line 2.0, Laboratory Chairs
: VWR Collection