Palladium standard solution, 1,000 mg/L Pd in 5% nitric acid standard for ICP
: CPACHEM
Frequently Bought Together
Buffer solutions in sachets
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
Niobium standard solution, 1000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for ICP, Specpure®
: Thermo Fisher Scientific