Scandium standard solution, 10,000 mg/L Sc in 5% nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
Triammonium citrate 97-103%, GPR RECTAPUR®
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals