Order Entry
Belgium
ContactUsLinkComponent
Iron standard solution, 100 mg/L Fe in dil. nitric acid standard for ICP-MS
Iron standard solution, 100 mg/L Fe in dil. nitric acid standard for ICP-MS
Catalog # ACSDM319.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Iron standard solution, 100 mg/L Fe in dil. nitric acid standard for ICP-MS
Catalog # ACSDM319.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item