Cerium standard solution, 10000 mg/L Ce in 5% nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
Colour Reference Solutions, Reag.Ph.Eur.
Supplier: REAGECON
Buffer solution, colour coded, ready to use
Supplier: Bernd Kraft
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
Supplier: YSI ENVIRONMENTAL
Gluconolactone 1 * 500 mg
Supplier: EHRENSTORFER
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
GLYCOPROTEIN PERFORMANCE TEST STANDARD 1 * 1 KIT
Supplier: WATERS
Cerium standard solution, 10000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
VWR®, Colour standards, Saybolt
Supplier: VWR Chemicals
4-Chloro-o-toluidine
Supplier: EHRENSTORFER
VWR®, Density standards referenced at 25 °C
Supplier: VWR Chemicals
Procymidone
Supplier: EHRENSTORFER
4,4´-DDE 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
Kadethrin certified 1 * 0,1 g
Supplier: EHRENSTORFER
TOLUENE D8 1 * 0,1 g
Supplier: EHRENSTORFER