Order Entry
Belgium
ContactUsLinkComponent
Calcium standard solution, 100 mg/L Ca in dil. nitric acid standard for ICP-MS
Calcium standard solution, 100 mg/L Ca in dil. nitric acid standard for ICP-MS
Catalog # ACSDM309.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Calcium standard solution, 100 mg/L Ca in dil. nitric acid standard for ICP-MS
Catalog # ACSDM309.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item