Cyclohexane ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAC0696-100ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
- Absorbance (210nm):max. 0.800
- Absorbance (220nm):max. 0.400
- Absorbance (230nm):max. 0.200
Specification Test Results
About this item
:
:84.16 g/mol
Merck Index:14,02723
Boiling Pt:81°C
Melting Pt:7 °C
Density:0,7785 g/cm³ (20 °C)
Flash Pt:-20°C
Flash Pt:–20°C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814