
Methyl orange ACS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:547-58-0
:
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
:327.34 g/mol
Merck Index:13,06127
Boiling Pt:~90 °C (1013 hPa)
Melting Pt:>300 °C
Density:∼1 g/cm³ (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:208-925-3
UN:3143
ADR:6.1,III
:GHS06
Specification Test Results
- Clarity of Solution:Pass
- Visual Transition Interval (pH 3.2 – 4.4):Pass
Specification Test Results
Frequently Bought Together