



Formic acid ≥98.0% (by GC) for LC-MS
Supplier: TCI
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Merck Index:14,04241
Boiling Pt:100°C
Melting Pt:8°C
Density:1,22 g/cm³ (25 °C)
Flash Pt:57°C
EINECS:200-579-1
UN:1779
ADR:8,II
REACH:01-2119491174-37
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
- Absorbance:to pass test
- Suitability for LC-MS analysis:to pass test
Specification Test Results
Frequently Bought Together