

Sodium hydroxide 2 mol/L in water
Catalog # TCIAS0543-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Concentration (Neutralization titration):1.9 to 2.1 mol/L
Specification Test Results
About this item
:
:40 g/mol
Merck Index:14,08627
Boiling Pt:109 °C (1013 hPa)
Density:Max, 1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
:GHS08
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548