Order Entry
Belgium
ContactUsLinkComponent
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98.0% (by titrimetric analysis) coupling agent for peptides synthesis
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98.0% (by titrimetric analysis) coupling agent for peptides synthesis
Supplier:  TCI
Certificates
undefined
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥98.0% (by titrimetric analysis) coupling agent for peptides synthesis
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together