L-Lysine ≥97.0% (by titrimetric analysis)
: TCI
:56-87-1
:
:C₆H₁₄N₂O₂
:MFCD00064433
:146.19 g/mol
:14,05636
:290 °C (1013 hPa)
:196 °C
:200-294-2
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:+24.0 to +27.0 deg (C=2, 5mol/L HCl)
- Solubility in Water:almost transparency
Frequently Bought Together