Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals