Yttrium standard solution, 1000 mg/L Y in 5% nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
N-Nitrosodiethylamine (NDEA), Reference Standard
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond