Sodium standard solution, 1000 mg/L Na in dil. nitric acid standard for ICP
: CPACHEM
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Calibration fluids and oils for refractometers
: BELLINGHAM STANLEY
VWR®, Conductivity standard solutions
: VWR Chemicals
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
: Thermo Fisher Scientific
NATRIUMACETAT TRIHYDRAT 1 * 2 g
: Merck
VWR®, Flow Cup viscosity standards
: VWR Chemicals
pH buffer solutions
: Hach
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
: Thermo Fisher Scientific