Lithium 1.000 mg/l dans l'acide nitrique dilué étalon mono-élément pour ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
[EN]LEAD 100 µG/ML STOCK SLN CHINESE PHA 1 * 100 mL
Supplier: REAGECON
Standards for water pollution QC check, sample demand, VHG
Supplier: LGC Standards PROMOCHEM
FENOXAPROP-P (R-ENANTIOMER) CERTIFIED 1 * 0,1 g
Supplier: EHRENSTORFER
Organic reference standards, canthaxanthine, Dr. Ehrenstorfer
Supplier: EHRENSTORFER
Riboflavine
Supplier: EDQM
Buffer solution, colour coded, ready to use
Supplier: Bernd Kraft
FONOFOS CERTIFIED 1 * 0,1 g
Supplier: EHRENSTORFER
[EN]HYDROCORTISONE IMPURITY F 1 * 100 mg
Supplier: Merck
PAH-MIX 9 100 NG/µL 1 * 10 mL
Supplier: EHRENSTORFER
Polyethylene Terephthalate, Reference Standards
Supplier: USP
Ammonium ion chromatography standards, VHG
Supplier: LGC Standards PROMOCHEM
Rifampin, Reference Standards
Supplier: USP
Supelco® TraceCERT® Organic Reference Standard, Fenamidone
Supplier: Merck