Nitrate standard solution, 1000 mg/L NO₃- in Water standard for ion chromatography
Supplier: CPACHEM
Certificates
Nitrate standard solution, 1000 mg/L NO₃- in Water standard for ion chromatography
Supplier: CPACHEM
About this item
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
D-(+)-Glucose, anhydrous, GPR RECTAPUR®
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck