Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Irritant
Environment
Zinc sulphate concentrate for dilution to 0.1 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4800.
Supplier:  Avantor
CAS Number:  
Corrosive
Irritant
Environment
Zinc sulphate concentrate for dilution to 0.1 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4800.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0998-0.1002

Specification Test Results

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302