Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate heptahydrate 99-102% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium sulphate heptahydrate 99-102% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Magnesium sulphate heptahydrate 99-102% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (MgSO₄7H₂O) (by EDTA titrn):
    99.0 - 102.0 %
  • Insoluble Matter:
    ≤ 0.005 %
  • pH of 5% Solution at 25°C:
    5.0 - 7.0
  • Nitrate (NO₃):
    ≤ 0.002 %
  • Ammonium (NH₄):
    ≤ 0.002 %
  • Calcium (Ca):
    ≤ 0.02 %
  • Potassium (K):
    ≤ 0.005 %
  • Sodium (Na):
    ≤ 0.005 %
  • Strontium (Sr):
    ≤ 0.005 %
  • Chloride (Cl):
    ≤ 5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 5 ppm
  • Trace Impurities - Manganese (Mn):
    ≤ 5.0 ppm

Specification Test Results

Related Information

Frequently Bought Together