Lead standard solution, 1000 mg/L Pb in dil. nitric acid standard for ICP
: CPACHEM
- Environmentally Preferable:
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
: VWR Chemicals
Potassium chromate 5% in aqueous solution Reag. Ph. Eur. 1069201
: VWR Chemicals