Barium standard solution, 10000 mg/L Ba in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR® Conductivity standard, CRM, 5000 μS/cm
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
BUFFER SOLUTION PH4 1 * 500 mL
Supplier: FF CHEMICALS
CHEM013 MEASUREMENT STANDARD
Supplier: MARKET SOURCE PART PROCESS
pH buffer, single-use, SINGLET™
Supplier: Hach