
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% (by HPLC, titration analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:536-46-9
:MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991
:209.12 g/mol
Merck Index:13,03280
Melting Pt:208°C
Storage Temperature:Refrigerator
EINECS:208-635-7
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 % (calcd.on dried substance)
- Drying loss:max. 1.0 %
- Suitability for sulfide-analysis:to pass test
Specification Test Results
Frequently Bought Together